Learn More
Aviva Systems Biology Corp Magee1 Antibody - C-terminal region (ARP57492_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57492_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Magee1. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: AI847422, DAMAGE, mMage-e1ApplicationWB. Species reactivity: Guinea Pig, Human, Mouse, Rat
Catalog No. 50-286-320