Learn More
Aviva Systems Biology Corp MAGEL2 Antibody - N-terminal region (ARP59891_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59891_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MAGEL2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human MAGEL2. Alternate names: NDNL1, nM15ApplicationWB. Species reactivity: Cow, Dog, Human, Pig, Rabbit
Catalog No. 50-286-868