Learn More
Bioworld Magenta 2-Way Test Tube Cap 500units
Catalog No. 50255182
Encompass
Made of polypropelene, provided non-sterile. For 25 mm diameter test tubes. Test tube cap that allows for gas exchange during plant culture. Prevents condensation from entering the tube. Filters incoming air and fits over various length test tubes. 500 units
Catalog No. 50-255-182 Supplier Bioworld Supplier No. 309300122
Made of polypropelene, provided non-sterile. For 25 mm diameter test tubes. Test tube cap that allows for gas exchange during plant culture. Prevents condensation from entering the tube. Filters incoming air and fits over various length test tubes. 500 units