Learn More
Bioworld Magenta B-Cap Jar Lid, 1000/cs
Catalog No. 50255174
Encompass
Used for plant tissue culture. Made of polypropylene, supplied non-sterile. Lid fits over baby food jars used in plant culture. Replaces items such as mylar film and rubber bands. The lid is autoclavable and reusable. Minimizes contamination. 1000/cs
Catalog No. 50-255-174 Supplier Bioworld Supplier No. 309300051
Used for plant tissue culture. Made of polypropylene, supplied non-sterile. Lid fits over baby food jars used in plant culture. Replaces items such as mylar film and rubber bands. The lid is autoclavable and reusable. Minimizes contamination. 1000/cs