Learn More
Bioworld Magenta GA-7 and GA-7-3 Vessel Lids 100 units
Catalog No. 50255177
Encompass
Magenta GA-7 and GA-7-3 Vessel Lids without Vents. on-vented lid for Magenta GA-7/ 7-3 Plant Culture Boxes (Cat# 765215 and 765217). Magenta lids are manufactured from polypropylene, to be both autoclavable and resuable. Sturdy and leak free. Dimensions: 77 x 77 mm. Sold as 100/pk
Catalog No. 50-255-177 Supplier Bioworld Supplier No. 309300081
Magenta GA-7 and GA-7-3 Vessel Lids without Vents. on-vented lid for Magenta GA-7/ 7-3 Plant Culture Boxes (Cat# 765215 and 765217). Magenta lids are manufactured from polypropylene, to be both autoclavable and resuable. Sturdy and leak free. Dimensions: 77 x 77 mm. Sold as 100/pk