Learn More
Bioworld Magenta GA-7 Plant Culture Box w/o Lid 100 units
Catalog No. 50255175
Encompass
Magenta GA-7 Plant Culture Boxes are manufactured from polycarbonate, to be both autoclavable and reusable. Non-sterile boxes are excellent for use in the plant culture protocols. Sturdy and leak free. The clear surface allows for sample viewing without disrupting growth. Boxes have a growing area of 6.125 sq. inches. Dimensions: 3" x 3" x 4". Sold as 100/cs
Catalog No. 50-255-175 Supplier Bioworld Supplier No. 309300061
Magenta GA-7 Plant Culture Boxes are manufactured from polycarbonate, to be both autoclavable and reusable. Non-sterile boxes are excellent for use in the plant culture protocols. Sturdy and leak free. The clear surface allows for sample viewing without disrupting growth. Boxes have a growing area of 6.125 sq. inches. Dimensions: 3" x 3" x 4". Sold as 100/cs