Learn More
Research Products International Corp Magenta-Gal [5-Bromo-6-chloro-3-indolyl-B-D-galactopyranoside], 100 Milligrams
SDP
Catalog No. 50488550
Encompass_Preferred
Chromogenic substrate of β-Galactosidase. Produce magenta color.
Catalog No. 50-488-550 Supplier Research Products International Corp Supplier No. B730000.1
Chromogenic substrate of β-Galactosidase. Produce magenta color.