Learn More
Alkali Scientific Magenta Gal [5 Bromo 6 chloro 3 indolyl β D galactopyranoside], 100 Mg
SDP
Catalog No. 5000660790
Encompass_Preferred
Magenta Gal (5 Bromo 6 chloro 3 indolyl β D galactopyranoside) is a chromogenic substrate designed for laboratory use in the detection of galactosidase activity. This 100 mg reagent is widely used in biochemical assays where the presence of β-galactosidase is of interest. The compound reacts with the enzyme to produce a colorimetric change, allowing for the identification and quantification of enzyme activity. It is commonly used in microbiological assays, gene expression studies, and diagnostic tests where the detection of β-galactosidase is required for understanding enzyme function and metabolic processes in cells and organisms.
Catalog No. 50-006-60790 Supplier Alkali Scientific Supplier No. AGE001CP
Magenta Gal (5 Bromo 6 chloro 3 indolyl β D galactopyranoside) is a chromogenic substrate designed for laboratory use in the detection of galactosidase activity. This 100 mg reagent is widely used in biochemical assays where the presence of β-galactosidase is of interest. The compound reacts with the enzyme to produce a colorimetric change, allowing for the identification and quantification of enzyme activity. It is commonly used in microbiological assays, gene expression studies, and diagnostic tests where the detection of β-galactosidase is required for understanding enzyme function and metabolic processes in cells and organisms.