Learn More
Abcam Magenta-Gal, Chromogenic beta-galactosidase substrate, 500MG
Catalog No. 502704670
Encompass_Preferred
MW 408.63 g/mol. Chromogenic substrate for β-galactosidase producing a red or magenta insoluble product. Used to determine the presence or absence of a cloned DNA insert in bacteria growing on agar plates. Designed to replace X-Gal in blue-white selection of recombinant bacterial colonies with the lac+ phenotype. Lac+ colonies grown in the presence of Magenta-Gal turn an intense red color, allowing for easy differentiation between lac+ and lac- colonies.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4670 Supplier Abcam Supplier No. AB146381500MG
MW 408.63 g/mol. Chromogenic substrate for β-galactosidase producing a red or magenta insoluble product. Used to determine the presence or absence of a cloned DNA insert in bacteria growing on agar plates. Designed to replace X-Gal in blue-white selection of recombinant bacterial colonies with the lac+ phenotype. Lac+ colonies grown in the presence of Magenta-Gal turn an intense red color, allowing for easy differentiation between lac+ and lac- colonies.
The product is subject to the following: Abcam Restricted Use Statement