Learn More
Alkali Scientific Magenta GlcA Cyclohexylammonium Salt [5 Bromo 6 chloro 3 indolyl β D glucuronic acid, cyclohexylammonium salt], 10 G
SDP
Catalog No. 5000660291
Encompass_Preferred
Magenta GlcA Cyclohexylammonium Salt (5 Bromo 6 chloro 3 indolyl β D glucuronic acid, cyclohexylammonium salt) is a chromogenic substrate used for the detection of glucuronic acid derivatives and β-glucuronidase activity. This 10 g reagent is ideal for large-scale assays and research applications that require precise detection of enzyme activity. When β-glucuronidase acts on the substrate, a colorimetric reaction occurs, making it easy to quantify enzyme activity. This substrate is widely used in clinical diagnostics, biochemical research, and studies on glucuronic acid metabolism, making it an essential tool for enzyme assays and metabolic analysis.
Catalog No. 50-006-60291 Supplier Alkali Scientific Supplier No. ENT003CP
Magenta GlcA Cyclohexylammonium Salt (5 Bromo 6 chloro 3 indolyl β D glucuronic acid, cyclohexylammonium salt) is a chromogenic substrate used for the detection of glucuronic acid derivatives and β-glucuronidase activity. This 10 g reagent is ideal for large-scale assays and research applications that require precise detection of enzyme activity. When β-glucuronidase acts on the substrate, a colorimetric reaction occurs, making it easy to quantify enzyme activity. This substrate is widely used in clinical diagnostics, biochemical research, and studies on glucuronic acid metabolism, making it an essential tool for enzyme assays and metabolic analysis.