Learn More
Bioworld Magenta Plant Culture 7-Way Tray 50/cs
Catalog No. 50255180
Encompass
Carries vessels or culture tubes through continuous plant tissue culture operations including washing, filling, autoclaving, transporting and incubating without extra handling. Shelving module, resting on incubation rack channels instead of shelves. Ventilates and eliminates hot spots between shelves, allows for more closely spaced shelves on rack. Allows more even lighting. Autoclavable and reusable. Capacity: 8 Magenta GA-7/ GA-7-3 or 36 25mm culture tubes. Dimensions: 6 x 12". Sold as 50/cs
Catalog No. 50-255-180 Supplier Bioworld Supplier No. 309300111
Carries vessels or culture tubes through continuous plant tissue culture operations including washing, filling, autoclaving, transporting and incubating without extra handling. Shelving module, resting on incubation rack channels instead of shelves. Ventilates and eliminates hot spots between shelves, allows for more closely spaced shelves on rack. Allows more even lighting. Autoclavable and reusable. Capacity: 8 Magenta GA-7/ GA-7-3 or 36 25mm culture tubes. Dimensions: 6 x 12". Sold as 50/cs