Learn More
Sigma Aldrich Fine Chemicals Biosciences MagentaTM 2-way cap natura
Catalog No. 502906098
Encompass
Cap with drip ring and groove for optional filterAutoclavable
Catalog No. 50-290-6098 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. C1934500EA
Cap with drip ring and groove for optional filterAutoclavable