Learn More
World Precision Instrument M1L Magnetic Stand
SDP
Supplier:  World Precision Instrument M1L
Encompass
The base of each stand exerts a powerful magnetic force that holds it solidly on ferrous metal surfaces - even vertically or upside down
This is the same magnetic base and support arm as M1, but it is equipped with a longer (14-inch) vertical post. These are an ideal manipulator stands.
Optional M5 10 mm clamp should be added for use with M3301, DC3001, MD4 and MMJ manipulators.
Catalog No. 50-822-965