Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Centrifuge/Stirrer and High Speed Mini Centrifuge Replacement Power Adapter

Catalog No. 17100451
Change view
Click to view available options
For Use With (Equipment):
Magfuge Centrifuge/Stirrer
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
17-100-451 Magfuge Centrifuge/Stirrer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-100-451 Supplier Fisherbrand™ Supplier No. 120527
Only null left
Add to Cart
Add to Cart

Replacement Power Adapter for use with Heathrow Scientific™ MagFuge™ and Gusto™, and compatible with Thermo Scientific™ MySpin 12™

  • Fisherbrand™ Centrifuge/Stirrer and High Speed Mini Centrifuge Replacement Power Adapter
  • Also for use with Heathrow Scientific™ MagFuge™ and Gusto™
  • Compatible with Thermo Scientific™ MySpin 12™

Specifications

Type Power Adapter
Material Plastic
For Use With (Equipment) Magfuge Centrifuge/Stirrer
Disposable Reusable
Color Black
Autoclavable Not Autoclavable
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.