Learn More
Thomas Scientific MAGic ClampT Clamp for 1 x microplate for H1000-MR, max. 4
SDP
Catalog No. 501460018
Encompass_Preferred
For use with Incu-Shaker Series and Orbi-Shaker Series The convenient MAGic Clamp™ System uses a unique, magnetic attachment method. It is an easy way to instantly change between flask clamps of different sizes.
Catalog No. 50-146-0018 Supplier Thomas Scientific Supplier No. 1201R55
For use with Incu-Shaker Series and Orbi-Shaker Series The convenient MAGic Clamp™ System uses a unique, magnetic attachment method. It is an easy way to instantly change between flask clamps of different sizes.