Learn More
Research Products International Corp MAGIC CLAMP PLAT UNIV 18X18
SDP
Catalog No. NC2099782
Encompass
MAGic Clamp Universal Platform is for use with Incu-Shaker Mini and Incu-Shaker 10L model Shaking Incubators. Shaker platform secures magnetic flask clamps or test tube trays (sold separately) during incubated shaking applications. Designed to fit Digital Incubator Shaker, 10L - SKU 248578.
This item is not returnable. View return policy
Catalog No. NC2099782 Supplier Research Products International Corp Supplier No. 248582
MAGic Clamp Universal Platform is for use with Incu-Shaker Mini and Incu-Shaker 10L model Shaking Incubators. Shaker platform secures magnetic flask clamps or test tube trays (sold separately) during incubated shaking applications. Designed to fit Digital Incubator Shaker, 10L - SKU 248578.