Learn More
Thomas Scientific MAGic Clamp platform for standard capacity shakers. (14 x 12")
SDP
Catalog No. 501461102
Encompass
High capacity, 24x125, 14x250, 9x500, 5x1L or 2x2L Digital, Touch-Screen control and accuracy Change and attach flask clamps is as little as one second with the unique MAGic Clamp platform The Orbi-Shaker ™ is the a compact orbital shaker with a powerful motor drive that can mix as many as five 1L flasks.  The convenient MAGic Clamp™ platform is also available for use with Erlenmeyer flasks and test tube racks.  The unique, magnetic attachment method is the easiest way to instantly change between flask clamps of different sizes. The powerful Orbi-Shaker is capable of mixing a 14lb load capacity up to speeds as high as 300 rpm. The standard platform (included) features a non-slip, rubber coated surface, ideal for tissue culture flasks, Petri dishes and staining trays. The convenient MAGic Clamp platform (patent pending) is also available for use with Erlenmeyer flasks and test tube racks. The unique, magnetic attachment method is the easiest way to instantly change
Catalog No. 50-146-1102 Supplier Thomas Scientific Supplier No. 1222U26
High capacity, 24x125, 14x250, 9x500, 5x1L or 2x2L Digital, Touch-Screen control and accuracy Change and attach flask clamps is as little as one second with the unique MAGic Clamp platform The Orbi-Shaker ™ is the a compact orbital shaker with a powerful motor drive that can mix as many as five 1L flasks.  The convenient MAGic Clamp™ platform is also available for use with Erlenmeyer flasks and test tube racks.  The unique, magnetic attachment method is the easiest way to instantly change between flask clamps of different sizes. The powerful Orbi-Shaker is capable of mixing a 14lb load capacity up to speeds as high as 300 rpm. The standard platform (included) features a non-slip, rubber coated surface, ideal for tissue culture flasks, Petri dishes and staining trays. The convenient MAGic Clamp platform (patent pending) is also available for use with Erlenmeyer flasks and test tube racks. The unique, magnetic attachment method is the easiest way to instantly change