Learn More
Thomas Scientific MAGic ClampT Tube Rack for H1000-MR, 32x15ml, max. 2
SDP
Catalog No. 501460019
Encompass_Preferred
For use with Incu-Shaker Series and Orbi-Shaker Series The convenient MAGic Clamp™ System uses a unique, magnetic attachment method. It is an easy way to instantly change between flask clamps of different sizes.
Catalog No. 50-146-0019 Supplier Thomas Scientific Supplier No. 1201R56
For use with Incu-Shaker Series and Orbi-Shaker Series The convenient MAGic Clamp™ System uses a unique, magnetic attachment method. It is an easy way to instantly change between flask clamps of different sizes.