Learn More
Thomas Scientific MAGic ClampT Tube Rack, 12x1.5ml horizontal, max. 2
SDP
Catalog No. 501457892
Encompass_Preferred
For use with Incu-Shaker Series and Orbi-Shaker Series The convenient MAGic Clamp™ System uses a unique, magnetic attachment method. It is an easy way to instantly change between flask clamps of different sizes.
Catalog No. 50-145-7892 Supplier Thomas Scientific Supplier No. 1201R58
Add to Cart
Add to Cart
For use with Incu-Shaker Series and Orbi-Shaker Series The convenient MAGic Clamp™ System uses a unique, magnetic attachment method. It is an easy way to instantly change between flask clamps of different sizes.