Learn More
Chemglass Life Sciences MAGic Clamp Universal Platform for flasks and tube racks (9.5 x 11.5)
SDP
Catalog No. 507533819
Encompass
MAGic Clamp Universal Platform for flasks and tube racks (9.5 x 11.5). For use with Incu-Shake shaking incubators.
Catalog No. 50-753-3819 Supplier Chemglass Life Sciences Supplier No. CLS2543003
MAGic Clamp Universal Platform for flasks and tube racks (9.5 x 11.5). For use with Incu-Shake shaking incubators.