Learn More
Thomas Scientific MAGic ClampT universal platform for high-capacity shakers (18 x 18")
SDP
Catalog No. 501460029
Encompass
For use with Incu-Shaker Series and Orbi-Shaker Series The convenient MAGic Clamp™ System uses a unique, magnetic attachment method. It is an easy way to instantly change between flask clamps of different sizes.
Catalog No. 50-146-0029 Supplier Thomas Scientific Supplier No. 1207X19
For use with Incu-Shaker Series and Orbi-Shaker Series The convenient MAGic Clamp™ System uses a unique, magnetic attachment method. It is an easy way to instantly change between flask clamps of different sizes.