Learn More
Sigma Aldrich Fine Chemicals Biosciences MAGic ClampTM magnetic cla1EA
Catalog No. 502900582
Encompass
The convenient MAGic Clam platform for use with Erlenmeyer flasks and test tube racks. The unique magnetic attachment method is the easiest way to instantly change between flask clamps of different sizes.designed to be used with Benchmark Scientific H1010 shaking incubators and OrbiShaker XL orbital shakers
Catalog No. 50-290-0582 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z7424391EA
The convenient MAGic Clam platform for use with Erlenmeyer flasks and test tube racks. The unique magnetic attachment method is the easiest way to instantly change between flask clamps of different sizes.designed to be used with Benchmark Scientific H1010 shaking incubators and OrbiShaker XL orbital shakers