Learn More
Immunochemistry Technology Magic Red™ Cathepsin B Assay Kit, 25 tests, Fluorescent, Cell-permeant, Non-cytotoxic
Catalog No. NC0576908
Encompass
The Magic Red™ Cathepsin B Assay Kit provides a reliable, non-cytotoxic solution for measuring intracellular cathepsin B activity in live cells. This assay utilizes a unique substrate that generates fluorescent signals upon enzyme cleavage, enabling real-time detection without the need for cell lysis. Ideal for various cell types and compatible across multiple species, this kit includes necessary components for complete analysis.
  • Features a Magic Red substrate for fluorescence-based detection
  • Detects cathepsin B activity with real-time analysis
  • Compatible with fluorescence microscopy and plate readers
  • Non-cytotoxic for live-cell applications
  • Includes Hoechst 33342 for nuclear staining
  • Fits a variety of cell types: suspension and adherent cells
  • Developed for research use only
  • Requires storage at +4°C for unopened kits
  • Suitable for mammalian, insect, and yeast cells
Catalog No. NC0576908 Supplier Immunochemistry Technology Supplier No. 937
The Magic Red™ Cathepsin B Assay Kit provides a reliable, non-cytotoxic solution for measuring intracellular cathepsin B activity in live cells. This assay utilizes a unique substrate that generates fluorescent signals upon enzyme cleavage, enabling real-time detection without the need for cell lysis. Ideal for various cell types and compatible across multiple species, this kit includes necessary components for complete analysis.
  • Features a Magic Red substrate for fluorescence-based detection
  • Detects cathepsin B activity with real-time analysis
  • Compatible with fluorescence microscopy and plate readers
  • Non-cytotoxic for live-cell applications
  • Includes Hoechst 33342 for nuclear staining
  • Fits a variety of cell types: suspension and adherent cells
  • Developed for research use only
  • Requires storage at +4°C for unopened kits
  • Suitable for mammalian, insect, and yeast cells