Learn More
Sigma Aldrich Fine Chemicals Biosciences Magic Touch 2TM mini ice p1EA
Catalog No. 502901376
Encompass
The new design of the Magic Touch 2 ice pans and ice buckets has improved durability and performance. With superior insulation properties and chemical resistance these ice pans can be used with ice dry ice dry ice-salt solvent slurries or liquid nitrogen. The non-sweating closed cell wont absorb moisture or odors and the pan is virtually unbreakable under normal use conditions.featuresSnug fitting lids for additional insulation and protectionExpanded urethane is lightweight for easy transportfill line indicatorsBuilt-in spouts to pour of unwanted liquid*Raised edges for ease in carryingLightly textured non-slip finishStackable*feature is not in 1.0L pans
Catalog No. 50-290-1376 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. BAM1680711011EA
The new design of the Magic Touch 2 ice pans and ice buckets has improved durability and performance. With superior insulation properties and chemical resistance these ice pans can be used with ice dry ice dry ice-salt solvent slurries or liquid nitrogen. The non-sweating closed cell wont absorb moisture or odors and the pan is virtually unbreakable under normal use conditions.featuresSnug fitting lids for additional insulation and protectionExpanded urethane is lightweight for easy transportfill line indicatorsBuilt-in spouts to pour of unwanted liquid*Raised edges for ease in carryingLightly textured non-slip finishStackable*feature is not in 1.0L pans