Learn More
Bel Art™ Magic Touch™ Icewares Hangs On Holders
Catalog No. 1167680
Change view
Click to view available options
Stainless-steel ice bucket pipettor holder
Stainless-steel ice bucket/pan tube holder
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
11-676-80 Stainless-steel ice bucket pipettor holder
11-676-81 Stainless-steel ice bucket/pan tube holder
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-676-80 Supplier Bel Art™ Supplier No. 6000/00
No more trying to locate supplies or fumbling to carry them. These handy racks slip on to the sides of ice buckets and ice pans to securely hold pipettors and test tubes, creating a portable workstation. Made of durable, stainless steel, Hangs On holders can be used almost anywhere - from cold rooms to chemical hoods - without rusting. Each Ice Bucket Pipettor Holder accommodates three single barrel pipettors. The Ice Bucket and Ice Pan Multitube Holder accepts three different sizes of conical or round-bottom tubes and four 1.5 to 2mL microcentrifuge tubes.
Order Info
For other packaging options, contact your Customer Service Representative.