Learn More
MSC Magid D-ROC Coated Gloves, Cut A2, Abrasion 6, 1 ea
Catalog No. 5000686263
Encompass
Magid GPD510 Hyperon® polyurethane palm coated work gloves provide comfortable ANSI cut level A2 hand protection for customers working with sharp materials. These gloves are shrink-wrapped for vending use, offering long-lasting protection and cost-savings.
  • Durable Hyperon® blended shell delivers breathability, comfort, and 360° cut protection.
  • Polyurethane palm coating maintains dexterity while delivering an excellent grip.
  • Affordable protection to meet the needs of any safety program.
  • Color-coded overcast acts as a visual aid for sizing and reduces fraying.
  • Machine washable for longer service life.
  • High puncture resistance.
Catalog No. 50-006-86263 Supplier MSC Supplier No. 70661533
Magid GPD510 Hyperon® polyurethane palm coated work gloves provide comfortable ANSI cut level A2 hand protection for customers working with sharp materials. These gloves are shrink-wrapped for vending use, offering long-lasting protection and cost-savings.
  • Durable Hyperon® blended shell delivers breathability, comfort, and 360° cut protection.
  • Polyurethane palm coating maintains dexterity while delivering an excellent grip.
  • Affordable protection to meet the needs of any safety program.
  • Color-coded overcast acts as a visual aid for sizing and reduces fraying.
  • Machine washable for longer service life.
  • High puncture resistance.