Hyperon® work gloves offer ultra-comfortable 360° ANSI level A2 cut protection for tasks involving sharp materials and fluids. The double-dipped three-quarter coating ensures long-lasting protection against liquids, oils, and solvents while maintaining dexterity.
- Ultra-lightweight, form-fitting 18-gauge Hyperon® shell provides bare hand dexterity, cool comfort, and 360° cut protection.
- Three-quarter undercoat protects against liquids, oils, and solvents.
- Double-dipped NitriX grip technology palm coating delivers rock-solid grip in oily applications, enhances abrasion resistance, and increases wear life.
- Dark color hides dirt and grime, extending worker wear time.
- Color-coded overcast aids in sizing and reduces fraying.
- Machine washable for longer service life.
- ANSI level A2 cut resistance and ANSI level 3 puncture resistance.