These lightweight 13-gauge work gloves feature DX Technology, an engineered yarn with strength-enhancing micro-particles. This eliminates heavy steel and fiberglass, making them 20% lighter than comparable HPPE gloves. Non-irritating and cool to the touch, they enhance worker comfort.
- DX Technology coreless construction without discomfort of fiberglass or steel
- Cool to the touch DX Technology enhances worker comfort
- Engineered 13-gauge yarn for extreme comfort, 20% lighter than traditional HPPE
- TriTek palm coating strengthens grip and minimizes saturation from heavy liquids and oil
- Hi-vis color improves worker protection in low-light conditions
- Shield graphic is color-coded by cut level for easy identification
- Color-coded overcast for sizing and reduced fraying
- Machine washable for extended service life