These D-ROC Chemical Resistant and Liquid-Proof Fully Coated Nitrile Work Gloves are designed for applications where sharp materials and fluids may be present, offering long-lasting protection against liquids, oils, and solvents with ANSI Level A3 cut protection.
- The 18-gauge Hyperon shell provides comfort and cut protection
- Hi-vis liner indicates chemical protection penetration
- Triple-dipped green nitrile full coat offers superior liquid and chemical resistance and maintains dexterity
- NitriX Grip Technology palm coating ensures grip in wet applications, enhanced abrasion resistance, and increased wear life
- Slip-on cuff with elastic gusset provides secure fit
- ANSI Level A3 Cut Resistance, ANSI Level 4 Abrasion Resistance, and ANSI Level 3 Puncture Resistance
- EN ISO 374-1:2016, Type B, JKL compliant