Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magid™ KnitMaster™ Half-Finger Machine Knit Nylon Glove Liners
SDP

Catalog No. 19045439
Change view
Click to view available options
Size:
Medium
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-045-439 Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-045-439 Supplier Magid™ Supplier No. 20NYM
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Form fitting, 100% nylon gloves are low-linting and feature a reversible design with a knit wrist that resists unraveling. Half-finger design increases dexterity for applications in the electronics/technology industries.

  • 100% nylon, seamless machine knit gloves
  • Long-lasting, reversible style with a knit wrist that resists unraveling
  • Half-finger design ensures a high level of dexterity
  • Color-coded overedge for easy size identification
  • Can be worn alone or used as a glove liner

Specifications

Construction Machine Knit
Coating Material Uncoated
Color White
Disposable Reusable
Hand Ambidextrous
Material Nylon
Size Medium
Sterility Non-sterile
Finger Style Half
For Use With (Application) Computer Manufacturing, Electronics, and Inspection
Glove Cuff Style Knit
Series KnitMaster™ 20NY
Length (English) 6.5 in.
Length (Metric) 16.5 in.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.