The Magid D-ROC GPD249 glove is a special purpose work glove designed for high heat, cut, and abrasion resistance. It features a 13-gauge Aramid shell and NitriX® Grip Technology palm coating, providing enhanced durability and dexterity for various applications.
- A 13-gauge Aramid shell provides dexterity and heat resistance
- Brushed acrylic lining enhances insulation
- NitriX® Grip Technology palm coating offers solid grip in oily conditions and abrasion resistance
- Nitrile dots on palm strengthen grip and increase heat resistance
- Nitrile-reinforced thumb saddle enhances durability
- Color-coded by cut level for easy identification
- Machine washable for longer service life
- ANSI Level 5 contact heat protection (up to 319°C)
- ANSI Level A2 cut, Level 4 puncture, and Level 4 abrasion resistance