Learn More
Sklar Instruments Magill Catheter Forceps, Sklar 6-1/2 in., Fenestrated, Straight, Serrated, Stainless Steel. EA.
SDP
Catalog No. 501181388 Shop All Sklar Instruments Products
Encompass_Preferred
Magill Catheter Forceps, Sklar. 6-1/2 in., Fenestrated, Straight, Serrated, Stainless Steel. Sklar Magill Catheter Forceps are used for grasping endotracheal tube during intubation and removing foreign bodies from the airway. They can also be used to grasp other types of catheters during insertion.
Catalog No. 50-118-1388 Supplier Sklar Instruments Supplier No. 071777
Magill Catheter Forceps, Sklar. 6-1/2 in., Fenestrated, Straight, Serrated, Stainless Steel. Sklar Magill Catheter Forceps are used for grasping endotracheal tube during intubation and removing foreign bodies from the airway. They can also be used to grasp other types of catheters during insertion.