Learn More
Med Vet International Miltex Vantage Magill Endotracheal Catheter Forceps, 9in., Adult Size
Catalog No. 502830184
Encompass_Preferred
The Miltex Vantage V92-70 Magill endotracheal catheter forceps are made from high quality Japanese stainless steel and measure 9" in length (adult size). The have a non-locking finger ring offset handle and serrated, fenestrated, oval tips.
Catalog No. 50-283-0184 Supplier Med Vet International Supplier No. MILV9270
The Miltex Vantage V92-70 Magill endotracheal catheter forceps are made from high quality Japanese stainless steel and measure 9" in length (adult size). The have a non-locking finger ring offset handle and serrated, fenestrated, oval tips.