Learn More
Med Vet International Miltex Vantage Magill Endotracheal Catheter Forceps, 7in., Child Size
Catalog No. 502829241
Encompass_Preferred
The Miltex Vantage V92-72 Magill endotracheal catheter forceps are made from high quality Japanese stainless steel and measure 9" in length (child size). The have a non-locking finger ring offset handle and serrated, fenestrated, oval tips.
Catalog No. 50-282-9241 Supplier Med Vet International Supplier No. MILV9272
The Miltex Vantage V92-72 Magill endotracheal catheter forceps are made from high quality Japanese stainless steel and measure 9" in length (child size). The have a non-locking finger ring offset handle and serrated, fenestrated, oval tips.