Learn More
Julabo USA Inc MAGIO MS-1000FF 230V -90C
SDP
Catalog No. 5000077843
Encompass_Preferred
Refrigerated heating circulator JULABO MAGIO MS-1000FF 208-230V/60Hz R290/R1270 Refrigerant -90 100 C 1 8kW heater Cooling 1 25kW at 20C 1 15kW at 0C 1 1kW at -20C 1kW at -40C 700W at -60C 200W at -80C Adjustable pressure-suction pump 16-31Lpm 3 5-13 3PSI Color touchscreen UI with USB RS232 RS485 Ethernet MODBUS communication USB data-logging Pt100 port multi-lingual programmable M16x1 pump fittings 8 and 12mm barbs 7 5-10L bath tank 21 7x 23 6 x 37 in 238lbs with locking casters
Catalog No. 50-000-77843 Supplier Julabo USA Inc Supplier No. 9032757.N1.13
Add to Cart
Add to Cart
Request a Quote
Refrigerated heating circulator JULABO MAGIO MS-1000FF 208-230V/60Hz R290/R1270 Refrigerant -90 100 C 1 8kW heater Cooling 1 25kW at 20C 1 15kW at 0C 1 1kW at -20C 1kW at -40C 700W at -60C 200W at -80C Adjustable pressure-suction pump 16-31Lpm 3 5-13 3PSI Color touchscreen UI with USB RS232 RS485 Ethernet MODBUS communication USB data-logging Pt100 port multi-lingual programmable M16x1 pump fittings 8 and 12mm barbs 7 5-10L bath tank 21 7x 23 6 x 37 in 238lbs with locking casters