Learn More
Cayman Chemical MAGL Substrate 1ea
SDP
Catalog No. 502896934
Encompass_Preferred
MAGL substrate has been tested and formulated to work exclusively with Cayman’s Monoacylglycerol Lipase Inhibitor Screening Assay Kit (Item No. 705192).
Catalog No. 50-289-6934 Supplier Cayman Chemical Supplier No. 7051931 EA
MAGL substrate has been tested and formulated to work exclusively with Cayman’s Monoacylglycerol Lipase Inhibitor Screening Assay Kit (Item No. 705192).