Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ MagMAX™ Cell and Tissue DNA Extraction Buffer

Catalog No. A45470
Encompass_Preferred
Change view
Click to view available options
No. of Reactions:
100 Preps
1000 Preps
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
A45470 1000 Preps
A45469 100 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A45470 Supplier Applied Biosystems™ Supplier No. A45470
  • $439.00
    $412.65 / Each of 1

    Save $26.35

    6% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

MagMAX DNA Cell and Tissue Extraction Buffer, a component of the MagMAX DNA Multi-Sample Ultra 2.0 Kit with Cell and Tissue Extraction Buffer, is offered separately here for those kit users who need additional buffer.

MagMAX DNA Cell and Tissue Extraction Buffer, a component of the MagMAX DNA Multi-Sample Ultra 2.0 Kit with Cell and Tissue Extraction Buffer, is offered separately here for those kit users who need additional buffer.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 600 mL Cell and Tissue DNA Extraction Buffer
Sample Type Cells, Tissue
For Use With (Application) Next-Generation Sequencing, PCR, Real-Time PCR
High-throughput Compatibility Automated Protocols, High-throughput Compatible, Manual Protocols
No. of Reactions 1000 Preps
Product Line MagMAX
Product Type Cell and Tissue DNA Extraction Buffer
Quantity 1000 Preps
Shipping Condition Room Temperature
Target Genomic DNA
Are there larger volume standalone reagent components available to scale up the MagMAX HMW DNA workflow?

Yes, the following larger volume standalone components are available for purchase separately:
- MagMAX HMW DNA Booster, 1,000 preps (1.0 mL) (Cat. No. A36625)
- MagMAX DNA Multi-Sample Ultra 2.0 Proteinase K (45 mL) (Cat. No. A36578)
- MagMAX DNA Multi-Sample Ultra 2.0 Binding Beads (45 mL) (Cat. No. A36579)
- MagMAX DNA Multi-Sample Ultra 2.0 Wash I Solution (1.1 L) (Cat. No. A36580)
- MagMAX DNA Multi-Sample Ultra 2.0 Lysis Binding Solution (450 mL) (Cat No. A36581)
- MagMAX DNA Multi-Sample Ultra 2.0 Elution Solution (120 mL) (Cat No. A36582)
- MagMAX DNA Multi-Sample Ultra 2.0 Enhancer Solution (45 mL) (Cat No. A36583)
- MagMAX Cell and Tissue DNA Extraction Buffer (600 mL) (Cat No. A45470)

If I have already purchased the MagMAX DNA Multi-Sample Ultra 2.0 Kit or standalone components, what do I need to extract high molecular weight DNA?

In addition to the MagMAX DNA Mult-Sample Ultra 2.0 product(s) you will need:
MagMAX HMW DNA Booster, 100 preps (Cat. No. A36624) or
In addition to the MagMAX DNA Mult-Sample Ultra 2.0 product(s) you will need:
MagMAX HMW DNA Booster, 100 preps (Cat. No. A36624) or MagMAX HMW DNA Booster, 1,000 preps (Cat No. A36625).

For cell and tissue samples include:
MagMAX Cell and Tissue DNA Extraction Buffer, 100 preps (Cat. No. A45469) or MagMAX Cell and Tissue DNA Extraction Buffer, 1,000 preps (Cat. No. A45470)
The MagMAX HMW DNA User Guide (MAN1001478)
The specific HMW DNA KingFisher automation protocol for your KingFisher Purification System (see product description page for download).
MagMAX HMW DNA Booster, 1,000 preps (Cat No. A36625).

For cell and tissue samples include:
MagMAX Cell and Tissue DNA Extraction Buffer, 100 preps (Cat. No. A45469) or MagMAX Cell and Tissue DNA Extraction Buffer, 1,000 preps (Cat. No. A45470)
The MagMAX HMW DNA User Guide (MAN1001478)
The specific HMW DNA KingFisher automation protocol for your KingFisher Purification System (see product description page for download).

Can the MagMAX HMW DNA Booster be paired with other genomic DNA isolation kits?

The MagMAX HMW DNA Booster is specifically designed and optimized for use with the MagMAX DNA Multi-Sample Ultra 2.0 (Cat. No. A36570) chemistry.

Can the MagMAX DNA Multi-Sample Ultra 2.0 Kit (Cat. No. A36570) be used to isolate genomic DNA from bone marrow aspirates and PBMCs?

Yes, we have protocols for using the MagMAX DNA Multi-Sample Ultra 2.0 Kit (Cat. No. A36570) to isolate genomic DNA from these sample types, but they require purchase of an additional Cell and Tissue Extraction Buffer. The kit and buffer can be purchased together as Cat. No. A45721 or the Cell and Tissue Extraction Buffer can be purchased as a stand alone item (Cat. No. A45469 or A45470).

The suggested workflow for bone marrow aspirates is as follows:
- Use only up to 200 µL of bone marrow aspirate
- Centrifuge at 200 x g for 10 mins
- Remove the supernatant (leaving about 50 µL of aspirate with the loose pellet) and resuspend the cell pellet in 400 µL of Cell and Tissue Extraction Buffer
- From this point, follow the protocol for isolation of DNA from buffy coat samples

The suggested workflow for 1-2.0 x 10E6 PBMCs is as follows:
- Centrifuge at 500-800 x g for 10 mins
- Remove the supernatant (leaving the cell pellet) an resuspend the cell pellet in 400 L of Cell and Tissue Extraction Buffer
- From this point, follow the protocol for isolation of DNA from cultured cell samples

Can the MagMAX DNA Multi-Sample Ultra 2.0 Kit (Cat. No. A36570) be used to isolate DNA from cultured cells and tissue?

Yes, we have protocols for using the MagMAX DNA Multi-Sample Ultra 2.0 Kit (Cat. No. A36570) to isolate DNA from cells and tissue, but they require purchase of an additional Cell and Tissue Extraction Buffer. The kit and buffer can be purchased together as Cat. No. A45721 or the Cell and Tissue Extraction Buffer can be purchased as a stand alone item (Cat. No. A45469 or A45470).

Do the MagMAX DNA Multi-Sample Ultra 2.0 cell and tissue DNA isolation protocols require an RNase A treatment step?

RNase A digestion is optional, but we highly recommend doing it. We recommend using PureLink RNase A (20 mg/mL) (Cat. No. 12091021).


For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.