Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ MagMAX™ Cell and Tissue DNA Extraction Buffer

Catalog No. p-7181495
Encompass_Preferred
Change view
Click to view available options
No. of Reactions:
100 Preps
1000 Preps
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
A45469 100 Preps
A45470 1000 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A45469 Supplier Applied Biosystems™ Supplier No. A45469
Only null left
Add to Cart
Edge
Add to Cart

MagMAX DNA Cell and Tissue Extraction Buffer, a component of the MagMAX DNA Multi-Sample Ultra 2.0 Kit with Cell and Tissue Extraction Buffer, is offered separately here for those kit users who need additional buffer.

MagMAX DNA Cell and Tissue Extraction Buffer, a component of the MagMAX DNA Multi-Sample Ultra 2.0 Kit with Cell and Tissue Extraction Buffer, is offered separately here for those kit users who need additional buffer.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 60 mL Cell and Tissue DNA Extraction Buffer
Sample Type Cells, Tissue
For Use With (Application) Next-Generation Sequencing, PCR, Real-Time PCR
High-throughput Compatibility Automated Protocols, High-throughput Compatible, Manual Protocols
No. of Reactions 100 Preps
Product Line MagMAX
Product Type Cell and Tissue DNA Extraction Buffer
Quantity 100 Preps
Shipping Condition Room Temperature
Target Genomic DNA

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.