Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ MagMAX™ CORE Nucleic Acid Purification Kits with Disruption Modules

Catalog No. A40011273
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 Reactions
500 Reactions
Includes:
MagMAX™ CORE Nucleic Acid Purification Kit, Disruption Plate, Without Clarifying Solution
MagMAX™ CORE Nucleic Acid Purification Kit, Disruption Plates Module, With Clarifying Solution
MagMAX™ CORE Nucleic Acid Purification Kit, Disruption Tube Module, With Clarifying Solution
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Includes
A40011273 500 Reactions MagMAX™ CORE Nucleic Acid Purification Kit, Disruption Plates Module, With Clarifying Solution
A40011271 500 Reactions MagMAX™ CORE Nucleic Acid Purification Kit, Disruption Plate, Without Clarifying Solution
A40011272 100 Reactions MagMAX™ CORE Nucleic Acid Purification Kit, Disruption Tube Module, With Clarifying Solution
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A40011273 Supplier Applied Biosystems™ Supplier No. A40011273
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MagMAX CORE™ Disruption solutions are intended to facilitate nucleic acid extraction. Flexible formats are designed to adapt to varying throughput needs and workflow requirements.

The Applied Biosystems MagMAX™ CORE™ Disruption Modules Family is intended to support the MagMAX™ CORE™ Nucleic Acid Purification Kit by providing flexible, modular options for sample preparation across a variety of research workflows.

These modules are designed to facilitate the processing of a broad range of sample types, including complex matrices such as feces, environmental samples, milk, and organ tissues. They can assist in the mechanical and/or chemical disruption of challenging sample materials to support downstream nucleic acid purification processes.

The Disruption Modules Family can be used in research applications involving organisms that may be difficult to lyse, such as Mycobacterium avium subsp. paratuberculosis (MAP).

Key features:

  • Convenience — Compatible with MagMAX™ CORE™ chemistry to streamline sample preparation
  • Confidence — Optimized extraction of nucleic acids from difficult-to-lyse samples
  • Flexible — 100-reaction disruption tube module for lower-throughput workflows or 500-reaction plate module for higher-throughput workflows
  • Ergonomy — Optimized workflow compared to previous mechanical lysis configurations.
  • Efficiency — High throughput workflow users can reduce single-tube consumption using the disruption plate option

Additional product details

Three configurations are available to accommodate different laboratory workflows, throughput needs, and application requirements.

MagMAX™ CORE Nucleic Acid Purification Kit with Disruption Tube Module (A40011272)

Designed for individual sample processing combined with a chemical pre-treatment:

  • MagMAX™ CORE Nucleic Acid Purification Kit (100 rxn)
  • Bead-beating tubes pre-filled with zirconia beads (x100)
  • Clarifying Solution bottle (200 mL)

MagMAX™ CORE Nucleic Acid Purification Kit with Disruption Plates Module (A40011273)

Designed for higher-throughput sample processing combined with a chemical pre-treatment:

  • MagMAX™ CORE Nucleic Acid Purification Kit (500 rxn)
  • 96-deep-well plates pre-filled with zirconia beads with adapted foil seals (x6)
  • Clarifying Solution bottle (1 L)

MagMAX™ CORE Nucleic Acid Purification Kit with Disruption Plates (A40011271)

Designed for high-throughput sample disruption workflows without chemical pre-treatment:

  • MagMAX™ CORE Nucleic Acid Purification Kit (500 rxn)
  • 96-deep-well plates pre-filled with zirconia beads with adapted foil seals (x6)

The MagMAX™ CORE kit and the MagMAX™ CORE Disruption Modules are designed to support streamlined laboratory workflows and convenient sample processing in research settings.

TRUSTED_SUSTAINABILITY

Specifications

Format Plate
Isolation Technology Magnetic Bead
Scale Maxi
For Use With (Application) Disruption, Extraction
Includes MagMAX™ CORE Nucleic Acid Purification Kit, Disruption Plates Module, With Clarifying Solution
Product Type Disruption Module
Quantity 500 Reactions

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.