Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ MagMAX™ DNA Multi-Sample Ultra 2.0 Proteinase K

Catalog No. A36578
Encompass_Preferred
Change view
Click to view available options
Quantity:
45 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A36578 45 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A36578 Supplier Applied Biosystems™ Supplier No. A36578
  • PRODUCT PROMOTION
    $1,260.00
    $1,064.00 / Each of 1

    Save $196.00

    16% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This Proteinase K solution is included in the MagMAX DNA Multi-Sample Ultra 2.0 Kit.

This Proteinase K solution is included in the MagMAX DNA Multi-Sample Ultra 2.0 Kit. It is available separately for applications that require more Proteinase K solution than is provided in the kit.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at room temperature.
Product Line MagMAX
Product Type DNA Multi-Sample Ultra 2.0 Proteinase K
Quantity 45 mL
Shipping Condition Room Temperature
Are there larger volume standalone reagent components available to scale up the MagMAX HMW DNA workflow?

Yes, the following larger volume standalone components are available for purchase separately:
- MagMAX HMW DNA Booster, 1,000 preps (1.0 mL) (Cat. No. A36625)
- MagMAX DNA Multi-Sample Ultra 2.0 Proteinase K (45 mL) (Cat. No. A36578)
- MagMAX DNA Multi-Sample Ultra 2.0 Binding Beads (45 mL) (Cat. No. A36579)
- MagMAX DNA Multi-Sample Ultra 2.0 Wash I Solution (1.1 L) (Cat. No. A36580)
- MagMAX DNA Multi-Sample Ultra 2.0 Lysis Binding Solution (450 mL) (Cat No. A36581)
- MagMAX DNA Multi-Sample Ultra 2.0 Elution Solution (120 mL) (Cat No. A36582)
- MagMAX DNA Multi-Sample Ultra 2.0 Enhancer Solution (45 mL) (Cat No. A36583)
- MagMAX Cell and Tissue DNA Extraction Buffer (600 mL) (Cat No. A45470)

If I have already purchased the MagMAX DNA Multi-Sample Ultra 2.0 Kit or standalone components, what do I need to extract high molecular weight DNA?

In addition to the MagMAX DNA Mult-Sample Ultra 2.0 product(s) you will need:
MagMAX HMW DNA Booster, 100 preps (Cat. No. A36624) or
In addition to the MagMAX DNA Mult-Sample Ultra 2.0 product(s) you will need:
MagMAX HMW DNA Booster, 100 preps (Cat. No. A36624) or MagMAX HMW DNA Booster, 1,000 preps (Cat No. A36625).

For cell and tissue samples include:
MagMAX Cell and Tissue DNA Extraction Buffer, 100 preps (Cat. No. A45469) or MagMAX Cell and Tissue DNA Extraction Buffer, 1,000 preps (Cat. No. A45470)
The MagMAX HMW DNA User Guide (MAN1001478)
The specific HMW DNA KingFisher automation protocol for your KingFisher Purification System (see product description page for download).
MagMAX HMW DNA Booster, 1,000 preps (Cat No. A36625).

For cell and tissue samples include:
MagMAX Cell and Tissue DNA Extraction Buffer, 100 preps (Cat. No. A45469) or MagMAX Cell and Tissue DNA Extraction Buffer, 1,000 preps (Cat. No. A45470)
The MagMAX HMW DNA User Guide (MAN1001478)
The specific HMW DNA KingFisher automation protocol for your KingFisher Purification System (see product description page for download).

Can the MagMAX HMW DNA Booster be paired with other genomic DNA isolation kits?

The MagMAX HMW DNA Booster is specifically designed and optimized for use with the MagMAX DNA Multi-Sample Ultra 2.0 (Cat. No. A36570) chemistry.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.