Learn More
Applied Biosystems™ MagMax™ Express-96 PCR Well Magnetic Head
Catalog No. 501147999
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 head
Catalog No. Quantity
50-114-7999 1 head
1 options
Catalog No. 50-114-7999 Supplier Applied Biosystems™ Supplier No. 4472991
Add to Cart
Add to Cart
Request a Quote
This is a magnetic head for the MagMAX™ Express-96 Deep Well Magnetic Particle Processor that is designed for use with PCR plates.
Includes one magnetic head. Not compatible with the KingFisher™ Flex 96 Magnetic Particle Processor.
TRUSTED_SUSTAINABILITY
Specifications
Shipping Condition Room Temperature
Product Line MagMAX
Quantity 1 head
Type Magnetic Head
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
For Research Use Only. Not for use in diagnostic procedures.