Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MagMAX™ Lysis/Binding Solution Concentrate

Catalog No. am8500
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AM8500 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AM8500 Supplier Invitrogen™ Supplier No. AM8500
Only null left
Add to Cart
Edge
Add to Cart

Invitrogen MagMAX Lysis/Binding Solution Concentrate is a component of all the MagMAX Isolation Kits that may be purchased separately.

Invitrogen MagMAX Lysis/Binding Solution Concentrate is a component of all the MagMAX Isolation Kits that may be purchased separately. Supplied in one 100 mL bottle.

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Buffer Lysis Buffer, Binding Buffer
Content And Storage Store MagMAX™ Lysis/Binding Solution Concentrate at room temperature.
Product Line Ambion, MagMAX
Quantity 100 mL
Shipping Condition Room Temperature
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.