Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ MagMAX™ Microbiome Bead Tubes

Catalog No. A42351
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 tubes
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A42351 100 tubes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A42351 Supplier Applied Biosystems™ Supplier No. A42351
  • PRODUCT PROMOTION
    $219.00
    $185.00 / Each of 1

    Save $34.00

    16% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

These bead tubes are included in the MagMAX Microbiome Ultra Nucleic Acid Isolation Kit.

These bead tubes are included in the MagMAX Microbiome Ultra Nucleic Acid Isolation Kit (Cat. No. A42358). They are made available separately for applications that require more tubes than are provided in the kit.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at room temperature.

Sample Type Bacteria, Blood, Buccal Samples, Cells, Fungi, Liquid Samples (e.g. Serum), Viral Samples, Yeast
High-throughput Compatibility High-throughput Compatible, Manual Protocols, Automated Protocols
For Use With (Application) Next-Generation Sequencing, RT-PCR (Endpoint), Real-Time PCR
Product Line MagMAX
Quantity 100 tubes
Shipping Condition Room Temperature
Target Viral DNA/RNA, Pathogen DNA/RNA, Total Nucleic Acids
How many beads are in each tube of the MagMAX Microbiome Bead Tubes?

The number of beads in each tube is proprietary.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.