Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ MagMAX™ Prime Viral/Pathogen Elution Buffer

Catalog No. A58148
Encompass_Preferred
Change view
Click to view available options
Quantity:
120 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A58148 120 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A58148 Supplier Applied Biosystems™ Supplier No. A58148
  • PRODUCT PROMOTION
    $598.00
    $505.00 / Each of 1

    Save $93.00

    16% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

MagMAX Prime Viral/Pathogen Elution Buffer is offered separately here for applications that require more Elution Buffer than is provided in the MagMAX Prime Viral/Pathogen NA Isolation kits.

MagMAX Prime Viral/Pathogen Elution Buffer is offered separately here for applications that require more Elution Buffer than is provided in the MagMAX Prime Viral/Pathogen NA Isolation kits.

TRUSTED_SUSTAINABILITY

Specifications

Bead Type Magnetic
Color Clear
Content And Storage Store at 15–25°C.
Format Bottle
Isolation Technology Beads
Sample Type Swabs in Transport Media, Saliva, Urine, Serum, Plasma
For Use With (Application) Next-Generation Sequencing, RT-PCR (Endpoint), Real-Time PCR
High-throughput Compatibility High-throughput Compatible, Automated Protocols
Key Functions Isolate and purify viral, bacterial, parasitic, and fungal nucleic acid
No. of Reactions Up to 2000 Reactions
Nucleic Acid Type DNA/RNA
Product Line MagMAX
Product Type Reagent
Quantity 120 mL
Sample Volume 200–400 μL
Shipping Condition Room Temperature
Target Viral, Bacterial, Parasitic, and Fungal Nucleic Acid
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.