Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ MagMAX™ Prime Viral/Pathogen NA Isolation Kit

Catalog No. A58145
Encompass_Preferred
Change view
Click to view available options
Quantity:
Up to 600 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A58145 Up to 600 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A58145 Supplier Applied Biosystems™ Supplier No. A58145
  • $1,254.00
    $1,203.65 / Each of 1

    Save $50.35

    4% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

The MagMAX Prime Viral/Pathogen NA Isolation Kit is a magnetic-bead-based nucleic acid purification kit for the isolation and purification of viral, bacterial, yeast, and fungal nucleic acid from biological specimens.

The MagMAX Prime Viral/Pathogen NA Isolation Kit is a magnetic-bead-based nucleic acid purification kit for the isolation and purification of viral, bacterial, parasitic, and fungal nucleic acid from human biological specimens.

Features of the MagMAX Prime Viral/Pathogen NA Isolation Kit include:

  • Designed to isolate and purify microbial nucleic acid from a range of biological samples such as swabs in transport media, saliva, stool, urine, plasma, and serum
  • Flexible workflow allowing various sample types to be batched on a single plate
  • Streamlined automation-ready protocols designed for basic and advanced lysis and advanced stool workflows
  • Fast automated KingFisher Flex, KingFisher Apex, KingFisher Apex Dx (RUO mode) scripts to allow for 96 samples to be processed in less than 60 minutes
  • Flexible protocol accommodates sample volume inputs of 200–400 μL
  • Elution volume of 200 μL for stool samples and 60 μL for all other sample types
  • Available lysis buffers and bead-beating accessories to extract nucleic acids from challenging organisms and raw/stabilized stool sample type

The kit is intended for use by qualified and trained clinical laboratory personnel specifically instructed and trained in automated magnetic bead purification techniques.

Compatible with Thermo Fisher infectious disease research assays

Have confidence that your nucleic acid extraction kit will work with your downstream application and enjoy a complete solution from Thermo Fisher Scientific.

Benefits of MagMAX magnetic bead–based purification

Magnetic beads offer many benefits compared to other technologies for isolating nucleic acid. Beads bind the nucleic acid more efficiently than glass-fiber filters, resulting in higher and more consistent yields. Additionally, because filters and vacuum manifolds are not used, there is no risk of filter clogging due to cellular particulates in samples. This clogging issue is of particular concern with protein-rich, large-volume samples such as whole blood or plasma that are commonly used for viral testing.

Accessories available for advanced workflow

Advanced lysis workflow

MagMAX Prime G+ Bacterial and Fungal Lysis Buffer (Cat. No. A59053)

Advanced stool workflow

MagMAX Prime Stool Lysis Buffer (Cat. No. A58154)

MagMAX Prime Bead Beating Tubes (Cat. No. A58155)

Standalone reagents

MagMAX Prime Viral/Pathogen Binding Beads (Cat. No. A58147)

MagMAX Prime Viral/Pathogen Elution Buffer (Cat. No. A58148)

MagMAX Prime Viral/Pathogen Wash I Solution (Cat. No. A58149)

MagMAX Prime Viral/Pathogen Binding Solution (Cat. No. A58150)

MagMAX Prime Viral/Pathogen Proteinase K (Cat. No. A58151)

MagMAX Prime Viral/Pathogen Proteinase K Dye (Cat. No. A58152)

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage

• 160 mL Binding Solution
• 300 mL Wash I Solution
• 6 mL Binding Beads
• 3 mL Proteinase K
• 36 mL Elution Buffer
• 30 μL Proteinase K dye

Store at 15–25°C.

Format Kit
Isolation Technology Magnetic Bead
Sample Type Swabs in Transport Media, Saliva, Urine, Serum, Plasma
For Use With (Application) Next-Generation Sequencing, RT-PCR (Endpoint), Real-Time PCR
High-throughput Compatibility High-throughput Compatible, Automated Protocols
Key Functions Isolate and purify viral, bacterial, parasitic, and fungal nucleic acid
No. of Reactions Up to 600 Reactions
Nucleic Acid Type DNA/RNA
Product Type Reagent
Quantity Up to 600 Reactions
Sample Volume 200–400 μL
Shipping Condition Room Temperature
Target Viral, Bacterial, Parasitic, and Fungal Nucleic Acid
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.