Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ MagMAX™ Pro HT NoSpin Plasmid MiniPrep Kit

Catalog No. A58310
Encompass_Preferred
Change view
Click to view available options
Product Type:
Automated, pDNA Extraction Kit
Quantity:
1152 Reactions
2304 Reactions
384 Reactions
96 Reactions
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Quantity
A58310 Automated, pDNA Extraction Kit 384 Reactions
A58309 Automated, pDNA Extraction Kit 96 Reactions
A58311 Automated, pDNA Extraction Kit 1152 Reactions
A58312 Automated, pDNA Extraction Kit 2304 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. A58310 Supplier Applied Biosystems™ Supplier No. A58310
Get Up to $3,000 of qPCR Products with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The MagMAX Pro HT NoSpin Plasmid Miniprep Kit enables scalable, ultra-rapid purification of high-quality, transfection-grade plasmid DNA (pDNA) from bacterial cultures. Using magnetic-bead technology, it simplifies workflows by eliminating centrifugation and vacuum filtration bottlenecks.

The Applied Biosystems MagMAX Pro HT NoSpin Plasmid Miniprep Kit provides a scalable and ultra-rapid approach for purifying high quality, transfection-grade plasmid DNA (pDNA) directly from bacterial culture. Utilizing advanced magnetic-bead technology, the MagMAX Pro HT NoSpin kit drastically simplifies and streamlines plasmid purification workflows by eliminating centrifugation and vacuum filtration bottlenecks.

MagMAX Pro HT NoSpin Plasmid Miniprep Kit features include:

  • Bead-based—utilizes dual para-magnetic beads for plasmid isolation without time-intensive processing steps (centrifugation, vacuum filtration)
  • Automation enabled—turn-key, automation-friendly protocols and scripts, allowing 96 samples to be processed in under 35 minutes on KingFisher Apex and KingFisher Flex systems
  • Fast plasmid DNA isolation—35 minutes from start to completion and ready for downstream applications
  • Wide range of compatibility—typical E. coli cultures, common medias, and range of plasmids all acceptable
  • High-quality pDNA yields of up to 20 μg, ready for downstream applications
  • Ready for transfection—isolated pDNA with very low endotoxin levels < 1 EU/μg

Specifically designed for high-throughput plasmid isolation, the MagMAX Pro HT NoSpin technology is automation friendly and suited for use with KingFisher Apex, KingFisher Flex, and KingFisher Presto purification systems, as well as integrated liquid handling robots.

The MagMAX Pro HT purification chemistry is both effective and ecofriendly. Kits include a novel endotoxin reduction buffer to remove pyrogenic contaminates, and buffers are free of ethanol and chaotropic salts.

pDNA purified with the MagMAX Pro HT NoSpin kit is suitable for a wide array of downstream applications, such as transfection, cloning, enzymatic restriction, in vitro transcription, and sequencing.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C BactBind Beads, PlasmidPure Beads, Endotoxin Reduction Buffer
15°C to 25°C RNAse A, Resuspension Buffer, Lysis Buffer, Precipitation Buffer, Wash 1 Buffer, Wash 2 Buffer, Elution Buffer
Format 96 Deep-well Plate
Isolation Technology Magnetic Bead
Scale Mini
Sample Type Bacteria, E. coli
Elution Volume 100 μL
For Use With (Equipment) KingFisher Apex, Flex and Presto Processors with 96 Deep Well Heads, Liquid Handling Robotic Systems
For Use With (Application) Cloning, Transfection, Protein Expression, Protein Production, mRNA production via In Vitro Transcription, Gene Editing, Sequencing, PCR, Nucleic acid labeling, PCR
Nucleic Acid Type Plasmid DNA (pDNA)
Plate Compatibility KingFisher 96 Deep Well Plates with or without barcodes
Product Type Automated, pDNA Extraction Kit
Quantity 384 Reactions
Purity Transfection Grade
Sample Volume Bacterial Culture, 1 mL per well
Test Time ~ 32 min. for 96 samples, 1 KingFisher 96 DWP
Yield Up to 20 μg pDNA using high copy plasmid
Size Distribution 5-12kb
Show More Show Less
Can I grow an overnight bacterial culture directly in the KingFisher 96-well plate when using the MagMAX Pro HT NoSpin Plasmid MiniPrep Kit?

Yes, the KingFisher 96-well plate can be used for overnight bacterial culture growth with the MagMAX Pro HT NoSpin Plasmid MiniPrep Kit. However, growth conditions depend on factors such as media type, host strain, plasmid copy number, and plasmid size. For best results, optimize orbital shaker settings and monitor OD600 to minimize well-to-well variation and ensure consistent plasmid yield.

How does plasmid copy number affect yields with the MagMAX™ Pro HT NoSpin Plasmid MiniPrep Kit?

High-copy plasmids yield higher amounts of DNA and are ideal for most downstream applications. Low-copy plasmids can be extracted, but yields may be lower and may not be sufficient for all transfection protocols. For low-copy plasmids, optimizing culture conditions, increasing culture volume, or performing multiple elution steps may help improve yield.

Which bacterial strains are compatible with the MagMAX Pro HT NoSpin Plasmid MiniPrep Kit?

The MagMAX Pro HT NoSpin Plasmid MiniPrep Kit is compatible with K12 E. coli derivatives, including DH5alpha, TOP10, and DH10B strains. Plasmid yield may vary with low-copy origins, strong host dependencies, or high genomic integration tendencies.

Should I be concerned about bead clumps in the BactBind Beads or PlasmidPure Beads bottles before use?

No, bead settling during transit is normal. This does not affect performance. We recommend vortexing well, before use.

What should I do if there are precipitates in the Lysis Buffer within the MagMAX Pro HT NoSpin Plasmid MiniPrep Kit?

We recommend warming the buffer at 30-37 degrees C for 30 min, and then inverting to mix until the precipitates dissolve.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.