Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ MagMAX™ Saliva gDNA Binding Beads

Catalog No. A39061
Encompass_Preferred
Change view
Click to view available options
Quantity:
45 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A39061 45 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A39061 Supplier Applied Biosystems™ Supplier No. A39061
Get Up to $3,000 of qPCR Products with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

MagMAX Saliva gDNA Binding Beads

These magnetic binding beads are a component of the MagMAX Saliva gDNA Isolation Kit.

These magnetic binding beads are a component of the MagMAX Saliva gDNA Isolation Kit. They are offered here as a stand-alone reagent for applications that require more magnetic binding beads than is provided in the kit.

TRUSTED_SUSTAINABILITY

Specifications

Isolation Technology Magnetic Bead
For Use With (Application) Next-Generation Sequencing, PCR
High-throughput Compatibility High-throughput Compatible, Manual Protocols, Automated Protocols
Product Line MagMAX
Product Type Saliva gDNA Binding Beads
Quantity 45 mL
Shipping Condition Room Temperature
Target Genomic DNA
Can I process less than 200 µL of saliva with the MagMAX Saliva gDNA Isolation Kit?

Yes you can, but the gDNA yields will be very low.

Will the MagMAX Saliva gDNA Isolation Kit work for sample types besides saliva?

No, this kit is specialized for freshly collected saliva and stabilized saliva samples only.

Do you sell the buffers and beads included in the MagMAX Saliva gDNA Isolation Kit as standalone items?

Yes, we offer the following kit components as stand-alone items to accommodate custom workflows or reagent volumes that have been optimized for your specific automation needs:
MagMAX Saliva gDNA Binding Beads: Cat. No. A39061
MagMAX Saliva gDNA Wash I Solution: Cat. No. A39062
MagMAX Saliva gDNA Lysis/Binding Solution: Cat. No. A39063
MagMAX Saliva gDNA Elution Solution: Cat. No. A39064

Can I use Tris-HCl or water to elute DNA from the MagMAX Saliva gDNA Binding Beads?

This is not recommended. Switching the elution solution will impact the maximum yield and may impact binding bead carry-over into the eluate.

With the MagMAX Saliva gDNA Isolation Kit, do I use different protocols for processing fresh and stabilized saliva?

No, the same protocols are used for fresh and stabilized saliva.

Do you offer the Wash II Solution from the MagMAX Saliva gDNA Isolation Kit as a standalone item?

Wash II Solution is 80% ethanol and we do not offer it as a stand-alone item. It must be purchased separately from a different vendor.

How much saliva sample can I use with the MagMAX Saliva gDNA Isolation Kit?

You can use 50-500 µL of saliva in a 96-well plate. We also have protocols for processing 1 mL and 2 mL of saliva in a 24-well plate.


For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.