Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Applied Biosystems™ MagMAX™ Total RNA Elution Buffer
Description
This elution buffer is included in the MagMAX-96 Total RNA Isolation Kit (Cat. No. AM1830) and MagMAX-96 Viral RNA Isolation Kit (AM1836). It is made available here for separate purchase.
Specifications
Specifications
| Content And Storage | Store at room temperature. |
| Product Line | MagMAX |
| Quantity | 10 mL |
| Shipping Condition | Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?