Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ MagMAX™ Viral/Pathogen Nucleic Acid Isolation Kit

Catalog No. p-9023010
Encompass_Preferred
Change view
Click to view available options
Quantity:
Up to 100 Preps
Up to 1000 Preps
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A42352 Up to 100 Preps
A48310 Up to 1000 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A42352 Supplier Applied Biosystems™ Supplier No. A42352
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The MagMAX Viral/Pathogen Nucleic Acid Isolation Kit is designed to recover RNA and DNA from virus and gram-negative bacteria in samples such as blood, swabs, urine, and viral transport media (VTM).

The MagMAX Viral/Pathogen Nucleic Acid Isolation Kit is designed to recover RNA and DNA from virus and gram-negative bacteria in samples such as blood, swabs, urine, and viral transport media (VTM). The kit utilizes MagMAX magnetic-bead technology, ensuring reproducible recovery of high-quality nucleic acid compatible with a broad range of applications, including real-time PCR, digital PCR, and next-generation sequencing.

Features of the MagMAX Viral/Pathogen Nucleic Acid Isolation Kit include:

  • Automation-ready protocols designed for the KingFisher Flex and KingFisher Duo Prime systems
  • Fast procedure allows for 96 samples to be processed in under 30 minutes
  • Flexible protocol accommodates sample volume inputs from 200 μL to 2 mL of biofluids or transfer media
  • No need for carrier RNA
  • Elution volumes ranging from 50 to 100 μL

Maximum sample input in a 96-deep well or 24-deep well plate format
The optimized reagents included in the MagMAX Viral/Pathogen kit allow you to maximize the amount of sample input, thereby increasing the amount of RNA and/or DNA recovered. Up to 400 μL of sample can be processed per well in a KingFisher 96-deep well plate. For larger sample volumes, protocols are available to process up to 2 mL of sample per well in a KingFisher 24-deep well plate.

Benefits of MagMAX magnetic bead–based purification
Magnetic beads offer many benefits compared to other technologies for isolating nucleic acid. Beads bind the nucleic acid more efficiently than glass-fiber filters, resulting in higher and more consistent yields. Additionally, because filters and vacuum manifolds are not used, there is no risk of filter clogging due to cellular particulates in samples. This clogging issue is of particular concern with protein-rich, large-volume samples such as whole blood or plasma that are commonly used for viral testing.

The Dynabeads MyOne Silane contained in the MagMAX Viral/Pathogen Total Nucleic Acid Isolation Kit have an optimized silica-like surface chemistry with a highly specific surface area that allows efficient kinetics and high sensitivity in nucleic acid capture. These aspects make Dynabeads MyOne Silane ideal for the recovery viral RNA and DNA present at low concentrations in body fluids. Once captured, these nucleic acids can then be eluted in 50 μL of elution buffer for use in downstream applications.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • 55 mL Binding Solution, store at room temperature
• 100 mL Wash Solution, store at room temperature
• 10 mL Elution Solution, store at room temperature
• 1 mL Proteinase K, store at room temperature
• 2 mL DNA/RNA Binding Beads, store at room temperature

Format 96 Deep-well Plate, 24 Deep-well Plate
Purification Time <30 min.
Product Line MagMAX
Quantity Up to 100 Preps
Shipping Condition Room Temperature
I accidentally stored my MagMAX Viral/Pathogen Nucleic Acid Isolation Kit at room temperature for several months. The beads are supposed to be stored at 4 degrees C. Are they still usable?

Yes, the kit is still usable.

I am getting MagMAX bead carryover in some wells of my plate. What might be causing this and how can I resolve it?

Some bead carryover is common and has not been documented to affect downstream assays. The most common cause of bead carryover is too much input sample. The best way to determine if the sample amount is the cause of the issue is to run PBS with Xeno (artificial DNA/RNA) in the lysis buffer and then check if there is still bead carryover. If that is not the cause, the initial sample may require special processing, such as bead beating or liquification. If there is still bead carryover without the original sample, and in particular, if the carryover is present in the same wells for multiple runs, then this may be an instrument alignment issue.

Do I need any additional consumables when using MagMAX kits on the MagMAX Express-96 instrument or a KingFisher instrument?

Plastic consumables need to be purchased separately in order to use the MagMAX reagents on the MagMAX Express-96 or a KingFisher instrument. The particular consumables and the amount required will vary by kit and protocol so please check the user guide for the particular MagMAX kit to be sure that all required plastic consumables are available prior to sample processing.

Where can I find Bindlt script protocol (.bdz) files for use of MagMAX kits with the KingFisher Flex Purification System and KingFisher Duo Prime Purification System?

The best place to download KingFisher Flex and KingFisher Duo Prime protocols for a particular MagMAX kit is the relevant MagMAX kit product page. The Bindlt script protocol (.bdz) files can be found under the Documents>Product literature section.

Can I scale up the MagMAX kit reaction volumes? If so, how should I do this?

Yes, reaction volumes for MagMAX kits can be scaled up. In general, the amount of MagMAX reagents should be scaled up proportionally for the increase in sample input amount. Please see the specific kit user guide for any recommendation. If none are included, then optimization may be necessary.

Will addition of BME (2-mercaptoethanol) to the MagMAX lysis buffer cause any issues with nucleic acid extraction?

Addition of BME (2-mercaptoethanol) to the lysis buffer of MagMAX kits is fine. This can be beneficial with more RNase-rich samples like certain types of tissues.
In general, we recommend adding 0.3 µL BME per 100 µL of lysate.

Which MagMAX kit do you recommend for extracting viral/bacterial RNA from swabs?

We recommend using the MagMAX Viral/Pathogen Nucleic Acid Isolation Kit (Cat. No. A42352) for extracting total nucleic acid from easily lysed viruses and Gram-negative bacteria. For difficult-to-lyse samples like Gram-positive bacteria and fungi, we recommend using the MagMAX Viral/Pathogen Ultra Kit (Cat. No. A42356).

How long is the protocol for the MagMAX Viral/Pathogen Nucleic Acid Isolation Kit?

It takes <40 min from sample to extracted nucleic acid for up to 96 samples.

What types of downstream analysis are compatible with the MagMAX Viral/Pathogen Nucleic Acid Isolation Kit and MagMAX Viral/Pathogen Ultra Nucleic Acid Isolation Kit?

The following downstream analysis are compatible with the MagMAX Viral/Pathogen Nucleic Acid Isolation Kit and MagMAX Viral/Pathogen Ultra Nucleic Acid Isolation Kit: PCR, qPCR, qRT-PCR, OpenArray analysis, microarray analysis, and next-generation sequencing.

What is the lower limit for viral titer with the MagMAX Viral/Pathogen Nucleic Acid Isolation Kit?

With the MagMAX Viral/Pathogen Nucleic Acid Isolation Kit, nucleic acid has been successfully detected from as low as 50 viral copies in the starting sample.

What types of pathogens are compatible with the MagMAX Viral/Pathogen Nucleic Acid Isolation Kit?

(-)ssRNA virus, (+)ssRNA virus, ssDNA virus, dsDNA virus, and Gram negative bacteria are compatible with the MagMAX Viral/Pathogen Nucleic Acid Isolation Kit.

What sample types are compatible with the MagMAX Viral/Pathogen Nucleic Acid Isolation Kit?

Human biofluid samples (e.g., blood, serum, plasma, urine, CSF, lavage, saliva), samples stored in transport media (e.g., BD Universal Viral Transport (UVT) Media), and bacteriophage culture are compatible with the MagMAX Viral/Pathogen Nucleic Acid Isolation Kit.

Are the MagMAX Viral/Pathogen and MagMAX Viral/Pathogen Ultra nucleic acid isolation kits automation-compatible?

The MagMAX Viral/Pathogen and MagMAX Viral/Pathogen Ultra nucleic acid isolation kits are compatible with all KingFisher instruments (Flex, Duo Prime, and Presto) and can also be used for manual processing in 96-well plates.

What is the volume input range for the MagMAX Viral/Pathogen Nucleic Acid Isolation Kit (Cat. No. A42352) and MagMAX Viral/Pathogen Ultra Nucleic Acid Isolation Kit (Cat. No. A42356)?

For the 96-deep well plate format: 200-400 µL sample volume input (200 µL for whole blood
For the 24-deep well plate format: 500-2,000 µL sample volume input (1,000 µL for whole blood)


For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.