Learn More
V&P Scientific Inc MAGNETIC PLATE, 96 Well, Standard or Deep Well, Round or Conical Bottom, 6 Bars, 50MGO
Catalog No. NC2378931
Encompass
MAGNETIC BEAD SEPARATION PLATE, 96 Well, Standard or Deep Well Plate, Round or Conical Bottom, 6 Bars, 50MGO, NdFeB Magnets, White Polycarbonate Magnet Frame, 1 Sided Pellet Location, Bar 3mm Wide, Axial - North Poles Up Magnetic Orientation, SLAS Footprint, Includes Registration Base
Catalog No. NC2378931 Supplier V&P Scientific Inc Supplier No. VP 771MCWZM1
MAGNETIC BEAD SEPARATION PLATE, 96 Well, Standard or Deep Well Plate, Round or Conical Bottom, 6 Bars, 50MGO, NdFeB Magnets, White Polycarbonate Magnet Frame, 1 Sided Pellet Location, Bar 3mm Wide, Axial - North Poles Up Magnetic Orientation, SLAS Footprint, Includes Registration Base